H-Phe-Met-Arg-Phe-NH₂ Acetate
Supplier: Frontier Scientific
4001566.0025 4001566.0100
H-2975.0025BAEA 261.76 USD InStock
H-2975.0025BA H-2975.0100BA
H-Phe-Met-Arg-Phe-NH₂ Acetate
Proteins and Peptides
A molluscan neuropeptide isolated from the ganglia of Macrocallista nimbosa. FMRF-amide seems to be involved in neuronal NO production in gastropoda. CAS Number (net): 64190-70-1.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty