(D-Lys³)-GHRP-6
Supplier: Frontier Scientific
4030692.0005 4030692.0025
H-3108.0005BAEA 117.38 USD InStock
H-3108.0005BA H-3108.0025BA
(D-Lys³)-GHRP-6
Proteins and Peptides
A ghrelin receptor antagonist, as well as of GHRP-6 (H-9990). Asakawa et al. found that peripherally administered (D-Lys³)-GHRP-6 (HwkWfK-amide) decreased food intake in lean mice, in mice with diet induced obesity, and in ob/ob obese mice, and that repeated administration of this GHS-R antagonist decreased body weight gain and improved glycaemic control in ob/ob obese mice.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty