Mouse;Rabbit;Rat (Cys¹⁸)-Atrial Natriuretic Factor (4-18) amide
Supplier: Frontier Scientific
4030384.0500 4030384.1000
H-3134.0500BAEA 506.38 USD InStock
H-3134.0500BA H-3134.1000BA
Mouse;Rabbit;Rat (Cys¹⁸)-Atrial Natriuretic Factor (4-18) amide
Proteins and Peptides
C-ANF 4-23 binds with high affinity to approximately 99% of ANF receptors in the isolated perfused rat kidney. It competes effectively with biologically active atrial peptides for binding sites but is devoid of agonist or antagonist action on the generation of cGMP in vascular smooth muscle and endothelial cells in culture.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty