Neuromedin B Trifluoroacetate
Supplier: Frontier Scientific
4020211.0001 4020211.0005 4020211.0025
H-3280.0001BAEA 134.03 USD InStock
H-3280.0001BA H-3280.0005BA H-3280.0025BA
Neuromedin B Trifluoroacetate
Proteins and Peptides
NMB, a bombesin-linke peptide, binds to the bombesin receptors BRS1-4. NMB is expressed in the pituitary, brain, pancreas, adrenal and gastrointestinal tract. Its expression in adipose tissue appears to be regulated by changes in energy balance. NMB could inhibit food intake and modulates stress-related behavior.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty