H-His-Cys-Lys-Phe-Trp-Trp-OH
Supplier: Frontier Scientific
4026848.0005 4026848.0025
H-3524.0005BAEA 200.96 USD InStock
H-3524.0005BA H-3524.0025BA
H-His-Cys-Lys-Phe-Trp-Trp-OH
Proteins and Peptides
HCKFWW, potent inhibitor of the HIV-encoded integrase (IN), the enzyme responsible for the integration of HIV DNA into the human genome. HCKFWW inhibits IN-mediated 3'processing and integration with an IC₅₀ of 2 µM. The peptide is also active on IN proteins from other retroviruses such as HIV-2, feline immunodeficiency virus and Moloney murine leukemia virus. It was also found to inhibit the disintegration reaction, suggesting that this hexapeptide acts at or near the catalytic site of IN.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty