Human;Mouse;Rat VIP Acetate
Supplier: Frontier Scientific
4011611.0001 4011611.0005
H-3775.0001BAEA 201.1 USD InStock
H-3775.0001BA H-3775.0005BA
Human;Mouse;Rat VIP Acetate
Proteins and Peptides
VIP, a 28-amino acid peptide belonging to the secretin-glucagon-CRF family, is widely distributed in the central and peripheral nervous systems. Acting both as a neurotransmitter and as a hormone, VIP is involved in a wide variety of biological activities, including vaso- and bronchodilation, smooth muscle relaxation, and stimulation of secretion. An injectable formulation of this peptide in combination with the adrenergic drug phentolamine is expected to provide a new and effective alternative for erectile dysfunction patients.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty