Bovine;Guinea pig;Human;Mouse;Pig;Rat GLP-1 (9-36) Amide Trifluoroacetate
Supplier: Frontier Scientific
4027697.0001 4027697.0005
H-4012.0001BAEA 725.27 USD InStock
H-4012.0001BA H-4012.0005BA
Bovine;Guinea pig;Human;Mouse;Pig;Rat GLP-1 (9-36) Amide Trifluoroacetate
Proteins and Peptides
For the long-acting GLP-1 analog liraglutide see H-6724.
Glucagon-like peptide-1 (GLP-1) is efficiently degraded by the enzyme dipeptidyl peptidase IV (DPP IV), yielding the major metabolite GLP-1 (9-36) amide. GLP-1 (9-36) amide does not affect glucose disposal in mice either in the presence or absence of intact GLP-1 receptors or in the presence or absence of stimulated insulin levels. This suggests that the GLP-1 metabolite is not involved in the regulation of glucose homeostasis. Elahi et al. have shown that this GLP-1 metabolite acts as a weak insulinotropic agent and inhibits hepatic glucose production.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty