Human Acetyl-Calpastatin
Supplier: Frontier Scientific
4027881.0500 4027881.1000
H-4076.0500BAEA 401.67 USD InStock
H-4076.0500BA H-4076.1000BA
Human Acetyl-Calpastatin
Proteins and Peptides
For additional calpain inhibitors, please see also our L-trans-Epoxysuccinyl Peptides.
Acetyl-calpastatin 27 peptide, AC27P, a calpain inhibitor homologous to the minimal inhibitory domain I of human calpastatin, was shown to modulate the amyloid β-protein 42 secretion ratio. This finding might point to the possibility of exploring compounds that inhibit Aβ42 secretion.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty