ACV Trifluoroacetate
Supplier: Frontier Scientific
4028307.0005 4028307.0025
H-4204.0005BAEA 791.51 USD InStock
H-4204.0005BA H-4204.0025BA
ACV Trifluoroacetate
Proteins and Peptides
Antimicrobial peptides are produced by plants and most organisms throughout the animal kingdom including humans. Antimicrobial peptides protect against a broad range of infectious agents, as bacteria, fungi, and viruses. The amphibian skin is an especially rich source of antimicrobial peptides. See also the product families: Hepcidins LL-37 and Fragments Tuftsin and Analogs (subfamily).
ACV is the key intermediate in the biosynthesis of all penicillin and cephalosporin antibiotics by eukaryotic and prokaryotic microorganisms.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty