H-Met-Ala-Ser-OH
Supplier: Frontier Scientific
4001423.0250 4001423.1000
H-4315.0250BAEA 163.38 USD InStock
H-4315.0250BA H-4315.1000BA
H-Met-Ala-Ser-OH
Proteins and Peptides
Substrate for human methionine aminopeptidases 1D and 2. For the latter enzyme, Yang et al. determined the kinetic parameters Km 0.38 ± 0.03 mM and kcat 210 ± 5 min⁻¹ at pH 7.5 and 30°C for the tripeptide MAS.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty