Human PAR-4 (1-6) Trifluoroacetate, Bachem
Supplier: Frontier Scientific
4028678.0005 4028678.0025
H-4348.0005BAEA 167.52 USD InStock
H-4348.0005BA H-4348.0025BA
Human PAR-4 (1-6) Trifluoroacetate, Bachem
Proteins and Peptides
PAR peptides (TRAP peptides and thrombin receptor-like peptides) activate the proteinase-activated receptors PAR-1 to PAR-4.
The hexapeptide GYPGQV corresponding to the amino terminus of the fourth protease-activated receptor (PAR4) after the cleavage at Arg⁴⁷/Gly⁴⁸ was tested for its ability to stimulate COS cells expressing PAR4. GYPGQV readily activated both wild-type and mutant PAR4 (R47A) at 500 µM, whereas thrombin and trypsin only activate the wild-type PAR4.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty