Human CART (55-102) Trifluoroacetate
Supplier: Frontier Scientific
4030508.0500 4030508.1000
H-4444.0100BAEA 536.66 USD InStock
H-4444.0100BA H-4444.0500BA H-4444.1000BA
Human CART (55-102) Trifluoroacetate
Proteins and Peptides
CART (Cocaine- and Amphetamine-Regulated Transcript) is a recently discovered peptide, which was initially identified as mRNA produced primarily in the rat hypothalamus after administration of psychomotor stimulants. It is a satiety factor and closely associated with the actions of two important regulators of food intake, leptin and neuropeptide Y. When systemic leptin or receptors for leptin are inhibited, expression of CART mRNA is suppressed. CART (55-102) has been isolated from the hypothalamus of ovine suggesting in vivo processing at the Lys⁵³-Arg⁵⁴ region. When injected intracerebroventricularly into rats, recombinant CART peptides inhibited both normal and starvation-induced feeding, and completely blocked the feeding response induced by neuropeptide Y. Thus far, CART (55-102) appears to be the most potent fragment with a conserved secondary structure consisting of three disulfide bridges. Disruption of this secondary structure by reduction leads to a loss of the appetite induction.
CART peptides, especially CART (55-102) appear to have an important function in the regulation of energy homeostasis. Intracerebroventricular administration of CART (55-102) reduces appetite and stimulates energy expenditure, whereas injection of the peptide into specific hypothalamic nuclei increases food intake.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty