Human;Rat CART (61-102) Trifluoroacetate
Supplier: Frontier Scientific
4030510.0500 4030510.1000
H-4448.0100BAEA 402.52 USD InStock
H-4448.0100BA H-4448.0500BA H-4448.1000BA
Human;Rat CART (61-102) Trifluoroacetate
Proteins and Peptides
CART (Cocaine- and Amphetamine-Regulated Transcript) is a recently discovered peptide, which was initially identified as mRNA produced primarily in the rat hypothalamus after administration of psychomotor stimulants. It is a satiety factor and closely associated with the actions of two important regulators of food intake, leptin and neuropeptide Y. When systemic leptin or receptors for leptin are inhibited, expression of CART mRNA is suppressed. CART (55-102) has been isolated from the hypothalamus of ovine suggesting in vivo processing at the Lys⁵³-Arg⁵⁴ region. When injected intracerebroventricularly into rats, recombinant CART peptides inhibited both normal and starvation-induced feeding, and completely blocked the feeding response induced by neuropeptide Y. Thus far, CART (55-102) appears to be the most potent fragment with a conserved secondary structure consisting of three disulfide bridges. Disruption of this secondary structure by reduction leads to a loss of the appetite induction.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty