Bovine;Human;Mouse;Rat Apelin-13
Supplier: Frontier Scientific
4029109.0001 4029109.0005
H-4566.0001BAEA 120.22 USD InStock
H-4566.0001BA H-4566.0005BA
Bovine;Human;Mouse;Rat Apelin-13
Proteins and Peptides
Apelin-13, generated from apelin-36, is an endogenous peptide ligand for the human APJ receptor, a putative receptor protein related to the angiotensin receptor (AT1). Apelin-13 exerted an acidification-rate-promoting activity in the CHO cells expressing the APJ receptor with a median effective concentration (EC₅₀) value of 0.37 nM. In a study of Valle et al. chronic central administration of apelin-13 over 10 days increased food intake, body weight, locomotor activity and body temperature in C57BL/6 mice.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty