Bovine;Human;Mouse;Rat (Pyr¹)-Apelin-13
Supplier: Frontier Scientific
4029110.0001 4029110.0005
H-4568.0001BAEA 206.12 USD InStock
H-4568.0001BA H-4568.0005BA
Bovine;Human;Mouse;Rat (Pyr¹)-Apelin-13
Proteins and Peptides
The pyroglutamyl form of apelin-13 exhibited a high activity with an EC₅₀ value of 0.30 nM. pERPRLSHKGPMPF is probably the final product of the post-translational modification of apelin and the biologically active endogenous ligand. (Pyr¹)-Apelin-13 has been identified as the predominant apelin isoform in the human heart. Pyr-Apelin-13 stimulates glucose utilization in normal and obese insulin-resistant mice. It could be required for the maintenance of insulin sensitivity.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty