Acetyl-(Cys³,Nle⁴,Arg⁵,D-2-Nal⁷,Cys¹¹)-alpha-MSH (3-11) Amide Trifluoroacetate
Supplier: Frontier Scientific
4029134.0001 4029134.0005
H-4598.0001BAEA 163.38 USD InStock
H-4598.0001BA H-4598.0005BA
Acetyl-(Cys³,Nle⁴,Arg⁵,D-2-Nal⁷,Cys¹¹)-alpha-MSH (3-11) Amide Trifluoroacetate
Proteins and Peptides
The cyclic MSH analog HS024 showed about 20-fold selectivity and very high affinity (Ki = 0.29 nM) for the melanocortin-4 (MC4) receptor and also increased the food intake, indicating that blockade of the MC4 receptor is a highly effective way to increase feeding.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty