Human PAR-2 (1-6) amide Trifluoroacetate, Bachem
Supplier: Frontier Scientific
4029152.0025 4029152.0100
H-4624.0025BAEA 401.95 USD InStock
H-4624.0025BA H-4624.0100BA
Human PAR-2 (1-6) amide Trifluoroacetate, Bachem
Proteins and Peptides
PAR peptides (TRAP peptides and thrombin receptor-like peptides) activate the proteinase-activated receptors PAR-1 to PAR-4.
SLIGKV-amide, which corresponds to the tethered ligand sequence of human PAR-2 (protease activated receptor 2), can be used to investigate receptor functions. For an inactive control see H-5882 and H-6428.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty