Polyphemusin II-Derived Peptide
Supplier: Frontier Scientific
4029155.0500 4029155.1000
H-4626.0500BAEA 653.59 USD InStock
H-4626.0500BA H-4626.1000BA
Polyphemusin II-Derived Peptide
Proteins and Peptides
Antimicrobial peptides are produced by plants and most organisms throughout the animal kingdom including humans. Antimicrobial peptides protect against a broad range of infectious agents, as bacteria, fungi, and viruses. The amphibian skin is an especially rich source of antimicrobial peptides. See also the product families: Hepcidins LL-37 and Fragments Tuftsin and Analogs (subfamily).
Showed the highest inhibitory activity against the T cell line-tropic HIV-1 infection through its specific binding to a chemokine receptor CXCR4 (IC₅₀ = 0.18 ± 0.041 nM) and the strongest inhibitory effect on the binding of an anti-CXCR4 monoclonal antibody to CXCR4 (IC₅₀ = 2.3 ± 0.20 nM).
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty