NFAT Inhibitor Trifluoroacetate
Supplier: Frontier Scientific
4034010.0500 4034010.1000
H-4874.0500BAEA 326.8 USD InStock
H-4874.0500BA H-4874.1000BA
NFAT Inhibitor Trifluoroacetate
Proteins and Peptides
The flow of information from calcium-mobilizing receptors to nuclear factor of activated T cells (NFAT)-dependent genes is critically dependent on interaction between the phosphatase calcineurin and the transcription factor NFAT. The high-affinity calcineurin-binding peptide potently inhibited NFAT activation and NFAT-dependent expression of endogenous cytokine genes in T cells, without affecting the expression of other cytokines that require calcineurin but not NFAT. NFAT inhibitors may be useful in treating chronic ailments such as myocardial hypertrophy, allergy, arthritis, and autoimmune disease.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty