TIP-39 Trifluoroacetate
Supplier: Frontier Scientific
4033516.0500 4033516.1000
H-4878.0500BAEA 515.34 USD InStock
H-4878.0500BA H-4878.1000BA
TIP-39 Trifluoroacetate
Proteins and Peptides
Tuberoinfundibular peptide, TIP-39, originally isolated from bovine hypothalamus was found to be a potent and selective agonist of the pTH2 receptor which regulates pituitary hormone secretion and spinal cord regions involved in pain perception. Synthetic TIP-39 activated the human and rat pTH2 receptors with EC₅₀ of 0.5 ± 0.12 nM and 0.8 ± 0.3 nM, respectively. TIP-39 was much more potent than pTH (EC₅₀ = 49 ± 23 nM) at the rat pTH2 receptor. The discovery of TIP-39 provides an important tool for the investigation of the pTH2 receptor functions inside and outside the nervous system.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty