(d(CH₂)₅¹,Tyr(Me)²,Orn⁸)-Oxytocin Trifluoroacetate
Supplier: Frontier Scientific
4034493.0001 4034493.0005
H-4928.0001BAEA 372.9 USD InStock
H-4928.0001BA H-4928.0005BA
(d(CH₂)₅¹,Tyr(Me)²,Orn⁸)-Oxytocin Trifluoroacetate
Proteins and Peptides
For atosiban see H-6722.
Potent oxytocin antagonist with a pA₂ value of 7.35 ± 0.08. (d(CH₂)₅¹,Tyr(Me)²,Orn⁸)-Vasotocin has been used to support the hypothesis that pituitary secretion of oxytocin is associated with coactivation of centrally projecting brain oxytocin pathways, some of which are causally related to the induced inhibition of food intake. Furthermore, this compound competitively inhibited arginine vasotocin (AVT) contractions without affecting AVT efficacy.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty