Mouse;Rat PAR-2 (1-6) amide Trifluoroacetate
Supplier: Frontier Scientific
4033483.0001 4033483.0005
H-5078.0001BAEA 140.98 USD InStock
H-5078.0001BA H-5078.0005BA
Mouse;Rat PAR-2 (1-6) amide Trifluoroacetate
Proteins and Peptides
PAR peptides (TRAP peptides and thrombin receptor-like peptides) activate the proteinase-activated receptors PAR-1 to PAR-4.
SLIGRL-amide, protease-activated receptor-2 (PAR2) selective agonist. For an inactive control see H-6508.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty