H-Val-His-Leu-Thr-Pro-OH
Supplier: Frontier Scientific
4010555.0005 4010555.0025
H-5275.0005BAEA 361.17 USD InStock
H-5275.0005BA H-5275.0025BA
H-Val-His-Leu-Thr-Pro-OH
Proteins and Peptides
N-terminal pentapeptide of human hemoglobin beta (Hb A and and the pathological HbS, which differ in position 6). Cellulose membrane-bound VHLTP was used as ligand for the anti-HbA1c antibody.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty