H-Val-Tyr-Val-OH
Supplier: Frontier Scientific
4001260.0250 4001260.1000
H-5300.0250BAEA 361.17 USD InStock
H-5300.0250BA H-5300.1000BA
H-Val-Tyr-Val-OH
Proteins and Peptides
VYV was used as internal standard peptide in urine analysis by LC-MS. The tripeptide is used in peptide standard mixtures for HPLC.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty