(d(CH₂)₅¹,Tyr(Me)²,Arg⁸)-Vasopressin Trifluoroacetate
Supplier: Frontier Scientific
4031349.0001 4031349.0005
H-5350.0001BAEA 171.56 USD InStock
H-5350.0001BA H-5350.0005BA
(d(CH₂)₅¹,Tyr(Me)²,Arg⁸)-Vasopressin Trifluoroacetate
Proteins and Peptides
For desmopressin see H-7675.
A very potent V₁ receptor antagonist (antivasopressor) with almost no antidiuretic activity.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty