Human Neuromedin U-25 Trifluoroacetate
Supplier: Frontier Scientific
4037521.0500 4037521.1000
H-5538.0500BAEA 326.8 USD InStock
H-5538.0500BA H-5538.1000BA
Human Neuromedin U-25 Trifluoroacetate
Proteins and Peptides
Neuromedin U is a neuropeptide which is widely distributed in the gut and central nervous system. Initially isolated from porcine spinal cord it was named for its uterine contractile activity. In fasted rats neuromedin U expression in certain brain areas is lower than in fed rats. In animal studies with neuromedin-23 (rat) overnight food intake is significantly reduced indicating that neuromedin U may be a physiological regulator of food intake. IC₅₀ values for the binding of neuromedin U-25 (human) to the human neuromedin U receptors NMU1R and NMU2R are 2.7 nM and 1.8 nM, respectively.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty