Human Kisspeptin-13 (4-13) Trifluoroacetate
Supplier: Frontier Scientific
4037534.0001 4037534.0005
H-5544.0001BAEA 189.51 USD InStock
H-5544.0001BA H-5544.0005BA
Human Kisspeptin-13 (4-13) Trifluoroacetate
Proteins and Peptides
Kisspeptin-10 (YNWNSFGLRF-amide), an LRF-amide motif-containing fragment of malignant melanoma metastasis suppressor KiSS-1, shows high agonistic potency towards the human G-protein-coupled receptor GPR54 also known as AXOR12. Kisspeptin-10 is a potent stimulator of LH and increases pulse frequency in men.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty