Mouse Recombinant Leptin
Supplier: Frontier Scientific
4038284.0200 4038284.1000
H-5582.0200BAEA 179.77 USD InStock
H-5582.0200BA H-5582.1000BA
Mouse Recombinant Leptin
Proteins and Peptides
Leptin, the product of the obese gene (ob), is a hormone secreted by adipocytes. Administration of recombinant mouse leptin in ob/ob mice - homozygous ob mutants suffering from obesity - corrected both overeating and obesity. Moreover leptin reduced the appetite and weight of mice with diet-induced obesity, and caused significant loss of body weight and fat content in lean mice. For other Leptin fragments please inquire.
16 kDa
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty