Human Intermedin Trifluoroacetate
Supplier: Frontier Scientific
4044529.0500 4044529.1000
H-6064.0500BAEA 670.32 USD InStock
H-6064.0500BA H-6064.1000BA
Human Intermedin Trifluoroacetate
Proteins and Peptides
Intermedins are members of the calcitonin/CGRP family. In vitro studies have shown that the synthetic 47 amino acid intermedin peptides act through the calcitonin receptor-like receptor/receptor activity-modifying protein (CRLR/RAMP) complexes by increasing intracellular cAMP levels. Unlike calcitonin gene-related peptide (CGRP) and adrenomedullin (ADM) intermedin exhibited no preference for one of the three RAMPs when co-expressed with CRLR. In normal and spontaneously hypertensive rats it has been demonstrated that intermedin treatment results in blood pressure reduction which can be blocked by the CGRP receptor antagonist CGRP (8-37). Apart from the hypotensive action, synthetic intermedin has also been shown to inhibit gastric emptying and food intake in mice. Therefore, intermedin represents an additional regulator of gastrointestinal and cardiovascular functions and might be involved in other bioactivities mediated by the CRLR/RAMP receptor complexes
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty