Rat Galanin-Like Peptide Trifluoroacetate
Supplier: Frontier Scientific
4047028.0500 4047028.1000
H-6204.0500BAEA 1307.18 USD InStock
H-6204.0500BA H-6204.1000BA
Rat Galanin-Like Peptide Trifluoroacetate
Proteins and Peptides
Galanin-like peptide (GALP), first isolated from the porcine hypothalamus and subsequently also identified in rats and humans, is a 60 amino acid neuropeptide that unlike galanin, has a non-amidated C-terminus. Its amino acid residues (9-21) are identical to the biologically active N-terminal (1-13) fragment of galanin. In contrast to galanin which is relatively non-selective, receptor binding studies revealed that GALP had a high affinity for the galanin receptor 2 (GALR2) and a lower affinity for the GALR1 receptor. Furthermore, it could be demonstrated that GALP stimulates food intake with a ten fold higher orexigenic activity than galanin and may also affect emotional state in the CNS.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty