Mouse;Rat (2-Furoyl)-PAR-2 (2-6)-Orn amide Trifluoroacetate
Supplier: Frontier Scientific
4048620.0005 4048620.0025
H-6246.0005BAEA 408.5 USD InStock
H-6246.0005BA H-6246.0025BA
Mouse;Rat (2-Furoyl)-PAR-2 (2-6)-Orn amide Trifluoroacetate
Proteins and Peptides
PAR peptides (TRAP peptides and thrombin receptor-like peptides) activate the proteinase-activated receptors PAR-1 to PAR-4.
2-(2-Furoyl)-PAR-2 (2-6)-Orn amide (mouse, rat) is a potent and selective PAR-2 agonist. In cultured human PAR-2-expressing cells (2-Furoyl)-LIGRLO-amide was equally effective as SLIGRL-amide (H-5078), in rat PAR-2-expressing cells it was 10 to 25 times more potent than SLIGRLamide in increasing intracellular calcium levels. In bioassays of tissue PAR-2 activity, measured as arterial vasodilation and hyperpolarization this agonist was 10 to 300 times more potent than H-5078.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty