H-Arg-Arg-Arg-Arg-Arg-Arg-Arg-Arg-Arg-OH Trifluoroacetate
Supplier: Frontier Scientific
4040952.0005 4040952.0025
H-6256.0005BAEA 374.62 USD InStock
H-6256.0005BA H-6256.0025BA
H-Arg-Arg-Arg-Arg-Arg-Arg-Arg-Arg-Arg-OH Trifluoroacetate
Proteins and Peptides
Nonaarginine is one of the most efficient protein transduction domains and a cell-penetrating peptide.
Sequence: H-Arg-Arg-Arg-Arg-Arg-Arg-Arg-Arg-Arg-OH
Salt: Trifluoroacetate
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty