Tyr-(D-Dab⁴,Arg⁵,D-Trp⁸)-Cyclo-Somatostatin-14 (4-11) Trifluoroacetate
Supplier: Frontier Scientific
4041674.0001 4041674.0005
H-6276.0001BAEA 502.75 USD InStock
H-6276.0001BA H-6276.0005BA
Tyr-(D-Dab⁴,Arg⁵,D-Trp⁸)-Cyclo-Somatostatin-14 (4-11) Trifluoroacetate
Proteins and Peptides
High affinity ligand for all five somatostatin receptors (sst1-sst5). In AtT-20 mouse anterior pituitary tumor cells which mainly express sst2 and sst5 receptors the binding affinity of this somatostatin analog was found to be higher than that of somatostatin-14 (H-1490) and octreotide (H-5972). In a model of epileptogenesis, however, KE 108 did not exhibit antiepileptic activity suggesting that the high potency and efficacy of a synthetic ligand to all known somatostatin receptors may not reproduce entirely the effects of the natural ligand.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty