H-Ser-Leu-Ser-Leu-Ser-Pro-Gly-OH Trifluoroacetate
Supplier: Frontier Scientific
4052978.0005 4052978.0025
H-6534.0005BAEA 653.59 USD InStock
H-6534.0005BA H-6534.0025BA
H-Ser-Leu-Ser-Leu-Ser-Pro-Gly-OH Trifluoroacetate
Proteins and Peptides
Peptide derived from the C-terminus of the human Ig Gamma-1 chain C region. SLSLPG was used for the structural characterization of recombinant hybrid IgG molecules by MS analysis of tryptic digests.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty