omega-Conotoxin GVIA Trifluoroacetate
Supplier: Frontier Scientific
4016306.0500 4016306.1000
H-6615.0500BAEA 502.75 USD InStock
H-6615.0500BA H-6615.1000BA
omega-Conotoxin GVIA Trifluoroacetate
Proteins and Peptides
ω-Conotoxin GVIA from the venom of the fish-hunting cone snail Conus geographus efficiently blocks neuronal calcium channels. This presynaptic inhibition of calcium influx prevents the voltage-activated release of acetylcholine.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty