Lepirudin Acetate (Disulfide Bonds, Random Oxidized)
Supplier: Frontier Scientific
4048773.0001 4048773.0005
H-6618.0001BAEA 2452.66 USD InStock
H-6618.0001BA H-6618.0005BA
Lepirudin Acetate (Disulfide Bonds, Random Oxidized)
Proteins and Peptides
These peptides correspond to the C-terminus of the 65-residue polypeptide hirudin, a potent thrombin inhibitor found in the saliva of the leech Hirudo medicinalis. Treatment of hirudin with carboxypeptidase Y has shown that the last 5 amino acids are essential for its inhibitory activity.
Synthetic peptide. Lepirudin is an anticoagulant which functions as a direct thrombin inhibitor. Offered under Bolar Exemption: All APIs that are sold for development of drug products still patent protected are offered under Bolar Exemption only. The following disclaimer applies: These products are offered and sold in small quantities only and solely for uses reasonably related to privileged trials and studies for obtaining marketing authorization required by law (Bolar Exemption). Bachem cannot be made liable for any infringement of intellectual property rights. It is the sole and only responsibility of the purchaser or user of these products to comply with the relevant national rules and regulations.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty