(Gly²²)-beta-Amyloid (1-40) Trifluoroacetate
Supplier: Frontier Scientific
4035372.0500 4035372.1000
H-6694.0500BAEA 474.73 USD InStock
H-6694.0500BA H-6694.1000BA
(Gly²²)-beta-Amyloid (1-40) Trifluoroacetate
Proteins and Peptides
The highly neurotoxic arctic mutant (E22G) of Aβ has been used to study the mechanisms underlying the formation of soluble and insoluble β-amyloid aggregates. As the wild-type Aβ, the arctic mutant preferably assembles in the presence of GM1 ganglioside.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty