beta-Amyloid (1-28) Trifluoroacetate
Supplier: Frontier Scientific
4025511.0500 4025511.1000
H-7865.0500BAEA 218.38 USD InStock
H-7865.0500BA H-7865.1000BA
beta-Amyloid (1-28) Trifluoroacetate
Proteins and Peptides
The remarkable histochemical features of Alzheimer's disease (AD) are plaques that contain cores of extracellular filaments, which have been identified as amyloid substance. The major component of this amyloid is the β-protein. The synthetic peptide, having the same sequence as the N-terminal 28 residues, formed fibrils that have the same antigenic and structural features like the native AD amyloid filaments.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty