Human Galanin Trifluoroacetate
Supplier: Frontier Scientific
4013896.0500 4013896.1000
H-8230.0500BAEA 496.15 USD InStock
H-8230.0500BA H-8230.1000BA
Human Galanin Trifluoroacetate
Proteins and Peptides
Human galanin, a 30-amino acid non-amidated peptide, has originally been isolated from colon and pituitary. Its sequence differs from that of rat and porcine galanins by 4 and 6 amino acids in the C-terminal moiety, respectively. Synthetic human galanin is equipotent to the rat peptide in the isolated rat fundus muscle strip bioassay. Both peptides also fully activate the rat galanin receptor, indicating the importance of the conserved N-terminal portion of galanin for receptor interaction. Galanin, which is widely distributed in the CNS, and GAL receptors (GALR) are overexpressed in limbic brain regions associated with cognition in Alzheimer disease. As galanin inhibits acetylcholine release, galanin antagonists have gained interest in AD research.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty