Human;Mouse;Pig;Rat;Sheep PACAP-38 Trifluoroacetate
Supplier: Frontier Scientific
4031157.0500 4031157.1000
H-8430.0200BAEA 353.77 USD InStock
H-8430.0200BA H-8430.0500BA H-8430.1000BA
Human;Mouse;Pig;Rat;Sheep PACAP-38 Trifluoroacetate
Proteins and Peptides
Kojro et al. observed that the PAC1 agonists PACAP-27 (H-1172) and PACAP-38 strongly increased the activity of α-secretase. Upregulation of this APP-degrading enzyme promotes the non-amyloidogenic processing of APP, i.e. reduces the production of Aβ40/42, and thus may help to prevent Alzheimer’s disease. Nasally applied PACAP-38 in APP[V717I]-transgenic mice additionally enhanced the production of the Aβ-degrading enzyme neprilysin via induction of somatostatin.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty