Z-D-Phe-Phe-Gly-OH
Supplier: Frontier Scientific
4015768.0250 4015768.1000
H-9430.0250BAEA 374.45 USD InStock
H-9430.0250BA H-9430.1000BA
Z-D-Phe-Phe-Gly-OH
Proteins and Peptides
A high level of inhibition of virus replication was obtained with this peptide, which resembles a region of a viral polypeptide.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty