(D-Trp⁷,Ala⁸,D-Phe¹⁰)-alpha-MSH (6-11) amide
Supplier: Frontier Scientific
4008401.0005 4008401.0025
H-9990.0005BAEA 163.38 USD InStock
H-9990.0005BA H-9990.0025BA
(D-Trp⁷,Ala⁸,D-Phe¹⁰)-alpha-MSH (6-11) amide
Proteins and Peptides
In search of competitive α-MSH antagonists, the hexapeptide GHRP-6 has been recognized as a selective inhibitor of α-MSH activity in the frog skin bioassay. It is identical in sequence with a growth hormone-releasing peptide analog, (His¹,Lys⁶)-GHRP, which selectively releases GH in vitro and in a wide variety of species in vivo. GHRP-6 also acts as a functional antagonist of somatostatin in the somatotrope cells.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty