Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Boc-Glu-OtBu

Supplier: Frontier Scientific

4000173.0001 4000173.0005 4000173.0025
A-1670.0001BAEA 114.5 USD InStock
A-1670.0001BA A-1670.0005BA A-1670.0025BA
Boc-Glu-OtBu
Boc-Glu-OtBu
Sequence: Boc-Glu-OtBu

The derivatives of Glu-NH₂ are assembled in the "Isoglutamine" subfamily.

Educt for obtaining a protected derivative of Glu γ-aldehyde used in the Fischer indole synthesis of Ar-substituted tryptophans.


Formula: C₁₄H₂₅NO₆
MW: 303.36 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00038273
CAS Number: 24277-39-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty