Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Boc-Nle-OH

Supplier: Frontier Scientific

4002807.0005 4002807.0025
A-2135.0005BAEA 179.77 USD InStock
A-2135.0005BA A-2135.0025BA
Boc-Nle-OH
Boc-Nle-OH
Sequence: Boc-Nle-OH (syrup)
Synonym(s): Boc-L-2-aminohexanoic acid (syrup)


Formula: C₁₁H₂₁NO₄
Storage Temperature: Freezer
MDL Number: MFCD00037270
CAS Number: 6404-28-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty