Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-(9-Fluorenylmethoxycarbonyl)-O-tert-butyl-L-threonine

Supplier: Frontier Scientific

4096755.0005 4096755.0025 4096755.0100
B-1245.0005BAEA 111.18 USD InStock
B-1245.0005BA B-1245.0025BA B-1245.0100BA
N-(9-Fluorenylmethoxycarbonyl)-O-tert-butyl-L-threonine
N-(9-Fluorenylmethoxycarbonyl)-O-tert-butyl-L-threonine
Sequence: Fmoc-Thr(tBu)-OH

Bachem also offers phosphothreonine derivatives.


Formula: C₂₃H₂₇NO₅
Storage Temperature: Refrigerator
MDL Number: MFCD00077075
CAS Number: 71989-35-0
EINECS: 276-261-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty