Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ac-Val-NH₂

Supplier: Frontier Scientific

4014244.0001 4014244.0005
E-1270.0001BAEA 114.5 USD InStock
E-1270.0001BA E-1270.0005BA
Ac-Val-NH₂
Ac-Val-NH₂
Sequence: Ac-Val-NH₂

Bachem additionally offers Fmoc-[¹⁵N]Val-OH, B-2670.


Formula: C₇H₁₄N₂O₂
Storage Temperature: Ambient
MDL Number: MFCD00080846
CAS Number: 37933-88-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty