Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

L-Cycloserine

Supplier: Frontier Scientific

4006516.0250 4006516.0050
F-1475.0250BAEA 603.32 USD InStock
F-1475.0250BA F-1475.0050BA
L-Cycloserine
L-Cycloserine
L-Cycloserine regulates sphingolipide metabolism by inhibiting the key enzyme serine palmitoyltransferase. Both cycloserine enantiomers inhibit 3-ketodihydrosphingosine synthetase (EC 2.3.1.50), though the L-form is the more potent inhibitor of the enzyme. Pérez-Sala et al. studied its inhibitory activity on pyruvate metabolism.


Formula: C₃H₆N₂O₂
MW: 102.09
Storage Temperature: Freezer
MDL Number: MFCD00064324
CAS Number: 339-72-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty