Abz-Gly-Ile-Val-Arg-Ala-Lys(Dnp)-OH
Supplier: Frontier Scientific
4049308.0001 4049308.0005
M-2595.0001BAEA 245.07 USD InStock
M-2595.0001BA M-2595.0005BA
Abz-Gly-Ile-Val-Arg-Ala-Lys(Dnp)-OH
Proteins and Peptides
Abz-GIVRAK(Dnp), a very efficient and selective FRET substrate for human cathepsin B (Km = 5.9 µM, kcat = 43 s⁻¹, kcat/Km = 7288 mM⁻¹s⁻¹). The kcat/Km values for cathepsin K, L, V, X, and cruzain were 133.3, 100, 32, 17, and 75 mM⁻¹s⁻¹, respectively.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty