1-Naphthalenylsulfonyl-Ile-Trp-aldehyde
Supplier: Frontier Scientific
4029154.0005 4029154.0025
N-1760.0005BAEA 555.54 USD InStock
N-1760.0005BA N-1760.0025BA
1-Naphthalenylsulfonyl-Ile-Trp-aldehyde
Proteins and Peptides
Please see also the inhibitors H-7650, N-1000, N-1040, N-1105, N-1125, N-1315, N-1320, N-1380, N-1395, and N-1855.
Cell-permeable and reversible inhibitor of cathepsin L, a lysosomal cysteine protease ( IC₅₀ 1.9 nM). 1-Naphthalenylsulfonyl-IW-CHO inhibited the release of Ca²⁺ and hydroxyproline from bone in an in vitro bone culture system. It should be useful for the treatment of osteoporosis.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty