Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Muscimol

Supplier: Frontier Scientific

4004216.0050 4004216.0250
Q-1345.0050BAEA 334.97 USD InStock
Q-1345.0050BA Q-1345.0250BA
Muscimol
Muscimol
Sequence: 5-(Aminomethyl)-3(2H)-isoxazolone
Synonym(s): Agarin#Pantherine

Muscimol, the major alkaloid in mushrooms of the Amanita family, is a potent, selective agonist of the GABA-A receptor. Also available in bulk quantities.


Formula: C₄H₆N₂O₂
Storage Temperature: Refrigerator
MDL Number: MFCD00057894
CAS Number: 2763-96-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty